Canine Recombinant NRG1/Neuregulin 1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
70084-D08H-100
103666-142EA 199.98 USD
103666-142
Canine Recombinant NRG1/Neuregulin 1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine NRG1 (XP_858147.1) extracellular domain (Ser19-Lys240) was expressed with a C-terminal polyhistidine tag.