Feline Recombinant IL1B/IL-1 beta (from E. coli)
Supplier: Sino Biological
67002-UNAE-20 67002-UNAE-100
76631-990EA 448 USD
76631-990 103668-940
Feline Recombinant IL1B/IL-1 beta (from E. coli)
Proteins and Peptides
A DNA sequence encoding the feline IL1B (NP_001070882.1) (Ala116-Ser267) was expressed with an initial Met.