Mouse Recombinant CD300LF Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
5A0540-M38H-50 5A0540-M38H-50
103886-476EA 179.78 USD
CA76476-646 103886-476
Mouse Recombinant CD300LF Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CD300LF (NP_001162624.1) (Met1-Ser193) was expressed with the Fc region of mouse IgG2a at the C-terminus.