Mouse I-TAC / CXCL11 protein (His Tag)
Supplier: Sino Biological
50151-M08Y-100 50151-M08Y-100
103887-804EA 351.48 USD
103887-804 CA76478-204
Mouse I-TAC / CXCL11 protein (His Tag)
Proteins and Peptides
A DNA sequence encoding the mouse Cxcl11 ( NP_062367.1) (Phe22-Met100) was expressed with a polyhistidine tag at the C-terminus.