Influenza A H3N2 (A/Amsterdam/4112/1992) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40742-V07H-100 40742-V07H-100
103887-484EA 401.98 USD
CA76477-884 103887-484
Influenza A H3N2 (A/Amsterdam/4112/1992) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the influenza A virus (A/Amsterdam/4112/1992 (H3N2)) neuraminidase (AFG72176) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.