Influenza A H5N1 (A/chicken/VietNam/NCVD-016/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40716-V08H-100 40716-V08H-100
103887-454EA 351.48 USD
103887-454 CA76477-854
Influenza A H5N1 (A/chicken/VietNam/NCVD-016/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/chicken/VietNam/NCVD-016/2008 (H5N1)) hemagglutinin (Met1-Ser526), termed as HA, was expressed with a polyhistidine tag at the C-terminus.