Influenza A H3N2 (A/swine/England/90591/1997) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40685-V08H-100 40685-V08H-100
103886-838EA 351.48 USD
103886-838 CA76477-238
Influenza A H3N2 (A/swine/England/90591/1997) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/swine/England/90591/1997 (H3N2)) hemagglutinin (AFR76481) (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.