Influenza A H1N1 (A/swine/OMS/2112/1995) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40660-V08H-100 40660-V08H-100
103887-368EA 351.48 USD
103887-368 CA76477-768
Influenza A H1N1 (A/swine/OMS/2112/1995) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/swine/OMS/2112/1995 (H1N1)) hemagglutinin (AFR76582)(Ala17-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.