Influenza A H5N8 (A/turkey/Germany-MV/R2472/2014 ) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40512-V08H1-100
103633-506EA 351.48 USD
103633-506
Influenza A H5N8 (A/turkey/Germany-MV/R2472/2014 ) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/turkey/Germany-MV/R2472/2014 (H5N8)) hemagglutinin (translated amino acids of EPI544756) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.