Influenza A H3N2 (A/Switzerland/9715293/2013) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40497-V08H1-100
103633-748EA 351.48 USD
103633-748
Influenza A H3N2 (A/Switzerland/9715293/2013) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Switzerland/9715293/2013(H3N2)) hemagglutinin (translated amino acids of EPI541659) (Met1-Arg345), termed as HA1, was expressed.