Influenza A H3N2 (A/Philippines/472/2002) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40487-V08B-100
103664-240EA 351.48 USD
103664-240
Influenza A H3N2 (A/Philippines/472/2002) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Philippines/472/2002 (MDCK)(H3N2)) hemagglutinin (AIU46063.1) (Met1-Trp530) was expressed with a polyhistidine tag at the C-terminus.