Influenza A H1N1 (A/Albany/12/1951) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40464-V08B-100
103664-190EA 351.48 USD
103664-190
Influenza A H1N1 (A/Albany/12/1951) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Albany/12/1951(H1N1)) hemagglutinin (ABP49481.1) (Met1-Arg344), termed as HA, was expressed with a polyhistidine tag at the C-terminus.