Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Recombinant GP2/Glycoprotein Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
40442-V04H-100
103664-140EA 351.48 USD
103664-140
Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Recombinant GP2/Glycoprotein Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Zaire ebolavirus(strain H.sapiens-wt/GIN/2014/Kissidougou-C15) GP (AHX24649.1) (Glu502-Gln650) was expressed with the Fc region of mouse IgG1 at the N-terminus.