Influenza A H5N8 (A/duck/Jiangsu/k1203/2010) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40385-V08H1-100
103634-018EA 351.48 USD
103634-018
Influenza A H5N8 (A/duck/Jiangsu/k1203/2010) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Jiangsu/k1203/2010(H5N8)) hemagglutinin (AFK13933.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.