Influenza A H5N8 (A/breeder duck/Korea/Gochang1/2014) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40383-V08H1-100
103633-978EA 351.48 USD
103633-978
Influenza A H5N8 (A/breeder duck/Korea/Gochang1/2014) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/breeder duck/Korea/Gochang1/2014 (H5N8)) hemagglutinin (AHL21419.1) (Met1-Lys341), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.