Influenza A H7N9 (A/Zhejiang/DTID-ZJU10/2013) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40325-V08B-100
103664-022EA 351.48 USD
103664-022
Influenza A H7N9 (A/Zhejiang/DTID-ZJU10/2013) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the influenza A virus (A/Zhejiang/DTID-ZJU10/2013(H7N9)) hemagglutinin (AHA11500.1) (Met1-Val524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.