Influenza A H9N2 (A/Hong Kong/3239/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40178-V08B-100
103663-904EA 351.48 USD
103663-904
Influenza A H9N2 (A/Hong Kong/3239/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hong Kong/3239/2008(H9N2) hemagglutinin (ADC41863.1) (Met1-Lys523), termed as HA, was expressed with a polyhistidine tag at the C-terminus.