Influenza A H3N2 (A/Hong Kong/1/1968) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40116-V08B-100
103663-758EA 351.48 USD
103663-758
Influenza A H3N2 (A/Hong Kong/1/1968) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hong Kong/1/1968(H3N2)) hemagglutinin (Q91MA7.1) (Met1-Trp530) was expressed with a C-terminal polyhistidine tag.