Influenza A H5N1 (A/Egypt/3300-NAMRU3/2008) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40049-V08H1-100
103632-786EA 351.48 USD
103632-786
Influenza A H5N1 (A/Egypt/3300-NAMRU3/2008) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the influenza A virus (A/Egypt/3300-NAMRU3/2008 (H5N1)) hemagglutinin (ACI06185.1) (Met1-Glu340), termed as HA1, was fused with a C-terminal polyhistidine tag.