Influenza A H4N4 (A/mallard duck/Alberta/299/1977) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40008-V08H-100
103663-476EA 351.48 USD
103663-476
Influenza A H4N4 (A/mallard duck/Alberta/299/1977) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Ile 528) of the influenza H4N4 hemagglutinin ((A/mallardduck/Alberta/299/1977 (H4N4)) (Q0A4G1), was fused with a polyhistidine tag at the C-terminus.