Human Recombinant Citrate synthase/CS Protein (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
14083-H08B-100
103623-588EA 351.48 USD
103623-588
Human Recombinant Citrate synthase/CS Protein (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human CS (O75390) (Met1-Gly466) was expressed with a C-terminal polyhistidine tag.