Human Recombinant GBA/glucocerebrosidase Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
12038-H08H-50 12038-H08H-50
103906-296EA 300.98 USD
103906-296 CA76479-956
Human Recombinant GBA/glucocerebrosidase Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human GBA (NP_000148.2) (Ala40-Gln536) was expressed with a polyhistidine tag at the C-terminus.