Influenza A H13N8 Recombinant (A/black-headed gull/Netherlands/1/00) Hemagglutinin/HA (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11721-V08H-100
103622-734EA 351.48 USD
103622-734
Influenza A H13N8 Recombinant (A/black-headed gull/Netherlands/1/00) Hemagglutinin/HA (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Lys 528) of the influenza A hemagglutinin H13N8 (A/black-headed gull/Netherlands/ 1/00 (H13N8)) (AAV91212.1), (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.