Influenza A H9N2 (A/Hong Kong/1073/99) Recombinant Hemagglutinin (HA1 Subunit) (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11229-V08H1-100
103633-864EA 351.48 USD
103633-864
Influenza A H9N2 (A/Hong Kong/1073/99) Recombinant Hemagglutinin (HA1 Subunit) (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hong Kong/1073/99(H9N2)) hemagglutinin (NP_859037.1) (Met1-Arg335), termed as HA1, was expressed with a C-terminal polyhistidine tag.