Human Recombinant TFRC/CD71 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11020-H01H-50 11020-H01H-50
103918-578EA 199.98 USD
CA76487-710 103918-578
Human Recombinant TFRC/CD71 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human TFRC (CAA25527.1) (Cys89-Phe760) was expressed with the Fc region of human IgG1 at the N-terminus.