Human Recombinant IL15RA & IL-15 (N120D) Heterodimer Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
CT094-H02H-100 CT094-H02H-100
103887-010EA 401.98 USD
103887-010 CA76477-410
Human Recombinant IL15RA & IL-15 (N120D) Heterodimer Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL15RA (NP_002180.1) (Ile31-Asp96) linked with the human IL15 (NP_000576.1, with mutation Asn 120 Asp) (Asn49-Ser162) by a peptide linker was fused with the Fc region of human IgG1 at the C-terminus.