Cynomolgus/Rhesus Recombinant PVRIG Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90971-C08H-100 90971-C08H-100 90971-C08H-1
103887-794EA 371.68 USD
103887-794 CA76478-194 76632-214
Cynomolgus/Rhesus Recombinant PVRIG Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus / rhesus PVRIG (XP_005549281.1/XP_014989941.2) (Thr41-Leu172) was expressed with a polyhistidine tag at the C-terminus.