Cynomolgus Recombinant Syndecan-1/CD138/SDC1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90938-C02H-200 90938-C02H-200
103886-984EA 452.48 USD
103886-984 CA76477-384
Cynomolgus Recombinant Syndecan-1/CD138/SDC1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus SDC1 (XP_005576533.1) (Met1-Glu251) was expressed with the Fc region of human IgG1 at the C-terminus.