Rhesus Recombinant Siglec-2/CD22 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90246-C08H-100
103585-478EA 351.48 USD
103585-478
Rhesus Recombinant Siglec-2/CD22 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus CD22 (EHH29920.1) (Met1-Arg686) was expressed with a polyhistidine tag at the C-terminus.