Rhesus Recombinant DDR2 Kinase/CD167b Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90228-C02H-100
103585-462EA 300.98 USD
103585-462
Rhesus Recombinant DDR2 Kinase/CD167b Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus DDR2 (XP_001118219.1) (Met1-Arg399) was expressed with the Fc region of human IgG1 at the C-terminus.