Cynomolgus Recombinant CD32b/FCGR2B Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90014-C08H-100 90014-C08H-1
103585-070EA 300.98 USD
103585-070 76632-600
Cynomolgus Recombinant CD32b/FCGR2B Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD32b (AAL92097.1) extracellular domain (Met 1-Pro 217) was fused with a polyhistidine tag at the C-terminus.