Human Recombinant CD157/BST1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10060-H08H-100 10060-H08H-50 10060-H08H-100
103620-748EA 300.98 USD
103620-748 CA103620-748 103678-258
Human Recombinant CD157/BST1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human BST1 (NP_004325.2) (Met 1-Lys 292) with a C-terminal polyhistidine tag was expressed.