Rat Recombinant B7-H3/CD276 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80380-R08H-100
103585-124EA 300.98 USD
103585-124
Rat Recombinant B7-H3/CD276 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat CD276 (Q7TPB4)(Met1-Phe244) was expressed with a polyhistidine tag at the C-terminus.