Rat Recombinant EpCAM/TROP-1/TACSTD1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80306-R02H-100
103666-612EA 300.98 USD
103666-612
Rat Recombinant EpCAM/TROP-1/TACSTD1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat EPCAM (O55159) (Met1-Thr266) was expressed, fused with the Fc region of human IgG1 at the C-terminus.