Rat Recombinant TNFRSF21/DR6 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80155-R02H-50
103669-400EA 179.78 USD
103669-400
Rat Recombinant TNFRSF21/DR6 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat TNFRSF21 (NP_001101677.1) extracellular domain (Met 1-His 349) was fused with the Fc region of human IgG1 at the C-terminus.