Human Recombinant TrkC/NTRK3 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10048-H08H-100 10048-H08H-50 10048-H08H-100
103678-150EA 191.35 USD
103678-150 103623-766 CA103623-766
Human Recombinant TrkC/NTRK3 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Asp 428) of human TrkC (NP_001007157.1) was expressed with a C-terminal polyhistide tag.