Mouse Recombinant SCF/C-kit ligand Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
50487-M08B-100
103665-066EA 401.98 USD
103665-066
Mouse Recombinant SCF/C-kit ligand Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the amino acids (Met 1-Ala 189) of mouse KITL (P20826-1) was fused with a polyhistidine tag at the C-terminus.