Mouse Recombinant CD28/TP44 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50103-M02H-100 50103-M02H-100
103886-436EA 199.98 USD
103886-436 CA76476-606
Mouse Recombinant CD28/TP44 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CD28 (NP_031668.3) (Met1-Lys149) was expressed with the Fc region of human IgG1 at the C-terminus.