Influenza A H1N1 (A/Brisbane/02/2018) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40719-V08H-100 40719-V08H-100
103918-758EA 351.48 USD
103918-758 CA76487-890
Influenza A H1N1 (A/Brisbane/02/2018) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Brisbane/02/2018 (H1N1)) hemagglutinin (translated amino acids of EPI1312566) (Met1-Ile530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.