Influenza A H1N1 (A/Texas/36/1991) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40713-V08H-100 40713-V08H-100
103887-450EA 351.48 USD
CA76477-850 103887-450
Influenza A H1N1 (A/Texas/36/1991) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Texas/36/1991 (H1N1)) hemagglutinin (ACF41933.1) (Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.