Influenza A H1N2 (A/Minnesota/14/2012) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40677-V08H-100 40677-V08H-100
103906-340EA 351.48 USD
CA76480-000 103906-340
Influenza A H1N2 (A/Minnesota/14/2012) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Minnesota/14/2012 (H1N2)) hemagglutinin (AHW83238) (Met1-Ser523), termed as HA, was expressed with a polyhistidine tag at the C-terminus.