Influenza A H1N2 (A/swine/Miyazaki/1/2006) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40672-V08H-100 40672-V08H-100
103887-392EA 351.48 USD
CA76477-792 103887-392
Influenza A H1N2 (A/swine/Miyazaki/1/2006) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/swine/Miyazaki/1/2006 (H1N2)) hemagglutinin (BAG49619)(Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.