Influenza A H3N2 (A/Hanoi/EL201/2009) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40490-V08H1-100
103633-742EA 351.48 USD
103633-742
Influenza A H3N2 (A/Hanoi/EL201/2009) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hanoi/EL201/2009 (H3N2)) hemagglutinin (AIU46093.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.