Influenza A H3N2 (A/Johannesburg/33/1994) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40476-V08B-100
103664-214EA 351.48 USD
103664-214
Influenza A H3N2 (A/Johannesburg/33/1994) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Johannesburg/33/1994)(H3N2)) hemagglutinin (AFM72164.1) (Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.