Influenza B (B/Wisconsin/01/2012) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40462-V08H1-100
103633-364EA 351.48 USD
103633-364
Influenza B (B/Wisconsin/01/2012) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza B virus (B/Wisconsin/01/2012) hemagglutinin (translated amino acids of EPI354025) (Met1-Arg362), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.