HIV-1 (group M, subtype B, strain SHIV-89.6P) Recombinant gp120 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40402-V08H-100
103663-646EA 351.48 USD
103663-646
HIV-1 (group M, subtype B, strain SHIV-89.6P) Recombinant gp120 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the HIV1(group M, subtype B, strain SHIV-89.6P) Envelope glycoprotein gp160 Protein(gp120 subunit) (AAC57427.1) (Thr31-Arg509) was expressed with a polyhistidine tag at the C-terminus.