Influenza A H3N2 (A/Victoria/3/1975) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40396-V08H1-100
103633-282EA 351.48 USD
103633-282
Influenza A H3N2 (A/Victoria/3/1975) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Victoria/3/1975(H3N2)) hemagglutinin (AFM71967.1) (Met1-Arg346), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.