Ebola virus EBOV (subtype Zaire, strain Mayinga 1976) Recombinant Glycoprotein/GP-RBD (Receptor Binding Domain) Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
40304-V02H1-100
103633-214EA 351.48 USD
103633-214
Ebola virus EBOV (subtype Zaire, strain Mayinga 1976) Recombinant Glycoprotein/GP-RBD (Receptor Binding Domain) Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Zaire ebolavirus (strain Mayinga 1976) GP (AAC54887.1) (Met1-Phe308) was expressed with the Fc region of human IgG1 at the C-terminus.