Influenza A H7N9 (A/Shanghai/2/2013) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40239-V08H-100
103663-962EA 351.48 USD
103663-962
Influenza A H7N9 (A/Shanghai/2/2013) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Shanghai/2/2013(H7N9)) hemagglutinin (Met1-Val524) was expressed with a C-terminal polyhistidine tag.