Influenza A H10N3 (A/mallard/Minnesota/Sg-00194/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40184-V08B-100
103663-918EA 351.48 USD
103663-918
Influenza A H10N3 (A/mallard/Minnesota/Sg-00194/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/mallard/Minnesota/Sg-00194/2007(H10N3)) hemagglutinin (ACT84107.1) (Met1-Val525) was expressed with a C-terminal polyhistidine tag.